Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002276.4 | NP_002267.2 | 5 | 1025 | ||
GenBank mRNA | AB041267.1 | - | 4 | 453 | ||
AB041268.1 | - | 4 | 452 | |||
AK313261.1 | - | 5 | 944 | |||
BC002539.2 | - | 5 | 929 | 3138504 | ||
BC007628.1 | - | 5 | 929 | 3504206 | ||
BC010409.2 | - | 5 | 936 | 3689178 | ||
BC067744.1 | - | 5 | 944 | 30332499 | ||
BC084574.1 | - | 5 | 927 | 6730698 | ||
Y00503.1 | - | 5 | 915 |