Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019990.4 | NP_064374.1 | 6 | 742 | ||
GenBank mRNA | AB031550.1 | - | 7 | 919 | ||
BC046335.1 | - | 6 | 831 | |||
BC058773.1 | - | 6 | 874 | |||
BC080808.1 | - | 6 | 937 | |||
BC094007.1 | - | 6 | 755 | |||
BC116984.1 | - | 5 | 650 | 40126303 | ||
BC120641.1 | - | 5 | 650 | 40129564 | ||
BG078855.2 | - | 5 | 528 |