Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019489.5 | NP_062362.1 | 6 | 337 | ||
GenBank mRNA | AF182825.1 | - | 6 | 319 | ||
AK008177.1 | - | 6 | 338 | |||
AK042165.1 | - | 3 | 2921 | |||
BC045154.1 | - | 6 | 332 | 6468589 |