Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207678.1 | NP_997561.1 | 2 | 360 | ||
GenBank mRNA | AB041605.1 | - | 2 | 337 | ||
AF211859.1 | - | 2 | 321 | |||
AK007552.1 | - | 2 | 357 | |||
AK008585.1 | - | 2 | 149 | |||
AK044204.1 | - | 2 | 297 | |||
AK048244.1 | - | 2 | 350 | |||
AK138938.1 | - | 2 | 71 | |||
AK156037.1 | - | 2 | 343 | |||
AK169857.1 | - | 2 | 336 | |||
AY337018.1 | - | 2 | 343 | |||
BC023747.1 | - | 2 | 335 | |||
BC132295.1 | - | 2 | 352 | 40130572 | ||
CD348864.1 | - | 2 | 360 |