Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207678.1 | NP_997561.1 | 3 | 460 | ||
GenBank mRNA | AB041605.1 | - | 3 | 437 | ||
AF211859.1 | - | 3 | 421 | |||
AK007552.1 | - | 3 | 457 | |||
AK008585.1 | - | 3 | 249 | |||
AK044204.1 | - | 3 | 397 | |||
AK048244.1 | - | 3 | 450 | |||
AK138938.1 | - | 3 | 171 | |||
AK156037.1 | - | 3 | 443 | |||
AK169857.1 | - | 3 | 436 | |||
AY337018.1 | - | 3 | 443 | |||
BC023747.1 | - | 3 | 435 | |||
BC132295.1 | - | 3 | 452 | 40130572 | ||
CD348864.1 | - | 3 | 460 |