Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rala | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019491.5 | NP_062364.3 | 3 | 464 | ||
GenBank mRNA | AF244951.1 | - | 2 | 227 | ||
AK013881.1 | - | 3 | 453 | |||
BC031741.1 | - | 3 | 464 | 4501311 | ||
Z48587.1 | - | 2 | 227 |