Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019494.1 | NP_062367.1 | 1, 2 | 133 | ||
GenBank mRNA | AF136449.1 | - | 1, 2 | 133 | ||
AF178672.1 | - | 1, 2 | 133 | |||
AF179872.1 | - | 1, 2 | 133 | |||
AK020750.1 | - | 1, 2 | 152 | |||
AK040051.1 | - | 1, 2 | 93 | |||
AK050012.1 | - | 2340 | ||||
BC025903.1 | - | 1, 2 | 121 | 5035983 |