Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcerg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039474.1 | NP_001034563.1 | 14 | 2104 | ||
NM_001289526.1 | NP_001276455.1 | 14 | 2092 | |||
GenBank mRNA | AB023485.1 | - | 14 | 2068 | ||
AK076993.1 | - | 14 | 2083 | |||
AK150312.1 | - | 14 | 2081 | |||
AK165067.1 | - | 1 | 62 | |||
AK168434.1 | - | 14 | 2082 | |||
BC039185.1 | - | 13 | 1994 | 3982736 | ||
BC040284.1 | - | 13 | 2004 | 4460736 |