Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019503.4 | NP_062376.2 | 4, 5 | 289 | ||
NM_052991.4 | NP_443717.1 | 4, 5 | 272 | |||
NM_052992.3 | NP_443718.1 | 4, 5 | 353 | |||
NM_194321.2 | NP_919302.1 | 4, 5 | 336 | |||
GenBank mRNA | AA462069.1 | - | 4, 5 | 221 | ||
AA466595.1 | - | 4, 5 | 289 | |||
AF089734.1 | - | 4, 5 | 215 | |||
AK002585.1 | - | 4, 5 | 276 | |||
BC024671.1 | - | 4, 5 | 226 | 4208962 | ||
BY064152.1 | - | 4, 5 | 288 | |||
BY132051.1 | - | 4, 5 | 258 | |||
CA459152.1 | - | 4, 5 | 294 | |||
CK793316.1 | - | 4, 5 | 271 | |||
W16239.1 | - | 4, 5 | 252 | |||
W98524.1 | - | 4, 5 | 267 |