Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019553.2 | NP_062426.2 | 9 | 1744 | ||
GenBank mRNA | AF159131.3 | - | 9 | 1705 | ||
AK010281.1 | - | 9 | 1624 | |||
AK145578.1 | - | 9 | 1728 | |||
AK160095.1 | - | 9 | 1718 | |||
BC030895.1 | - | 8 | 1176 | |||
BC043655.1 | - | 9 | 1716 | 5363948 | ||
BC059237.1 | - | 9 | 1704 | 6412782 | ||
BC060220.1 | - | 9 | 1696 |