Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019580.4 | NP_062526.1 | 2 | 440 | ||
GenBank mRNA | AF212860.1 | - | 2 | 421 | ||
AK004583.1 | - | 2 | 453 | |||
AK005361.1 | - | 2 | 441 | |||
AK150807.1 | - | 2 | 462 | |||
BC003902.1 | - | 2 | 399 | 3601420 |