Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019570.3 | NP_062516.2 | 3 | 320 | ||
GenBank mRNA | AB057418.1 | - | 2 | 109 | ||
AF179302.1 | - | 2 | 109 | |||
AK048838.1 | - | 3 | 325 | |||
BC058093.1 | - | 3 | 320 | 6827108 |