Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019564.3 | NP_062510.2 | 5, 6 | 1066 | ||
GenBank mRNA | AF172994.1 | - | 5, 6 | 1042 | ||
AF179369.1 | - | 5, 6 | 1037 | |||
AK090320.1 | - | 5, 6 | 781 | |||
AK090321.1 | - | 2, 3 | 213 | |||
AK128916.1 | - | 5, 6 | 1039 | |||
AK132270.1 | - | 5, 6 | 1039 | |||
AK167667.1 | - | 5, 6 | 632 | |||
BC013516.1 | - | 5, 6 | 1036 | 4235912 |