Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085472.2 | NP_001078941.2 | 7 | 687 | ||
NM_001085473.2 | NP_001078942.2 | 13 | 3012 | |||
NM_001242605.1 | NP_001229534.1 | 6 | 751 | |||
NM_001242606.1 | NP_001229535.1 | 6 | 712 | |||
NM_019567.3 | NP_062513.3 | 7 | 816 | |||
NM_023190.3 | NP_075679.2 | 13 | 3048 | |||
GenBank mRNA | AF124725.1 | - | 7 | 747 | ||
AF124729.1 | - | 6 | 682 | |||
AF168782.1 | - | 13 | 3049 | |||
AK012099.1 | - | 7 | 680 | |||
AK030449.1 | - | 13 | 2742 | |||
AK122342.1 | - | 13 | 2683 | |||
BC019127.1 | - | 6 | 736 | |||
BC052755.1 | - | 6 | 672 | |||
BC075691.1 | - | 13 | 2697 | |||
BC094217.1 | - | 6 | 694 | |||
BY711288.1 | - | 7 | 680 |