Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cited4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019563.2 | NP_062509.1 | 1 | 955 | ||
GenBank mRNA | AF143369.1 | - | 1 | 922 | ||
AK012136.1 | - | 1 | 947 | |||
AK154030.1 | - | 1 | 946 | |||
BC025116.1 | - | 2 | 627 | |||
BC080723.1 | - | 2 | 712 | |||
BC132360.1 | - | 1 | 902 | 40130637 | ||
BC132362.1 | - | 1 | 902 | 40130639 |