Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019571.5 | NP_062517.1 | 4 | 741 | ||
GenBank mRNA | AF121344.1 | - | 4 | 756 | ||
AK013350.1 | - | 4 | 706 | |||
AK015705.1 | - | 4 | 463 | |||
AK033543.1 | - | 4 | 440 | |||
AK187703.1 | - | 3 | 255 | |||
BC046784.1 | - | 4 | 644 | 6416107 | ||
BC058695.1 | - | 4 | 712 | 6813872 |