Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021299.1 | NP_067274.1 | 2 | 252 | ||
GenBank mRNA | AB020203.1 | - | 2 | 155 | ||
AK004864.2 | - | 2 | 267 | |||
AK005194.2 | - | 2 | 306 | |||
AK007618.1 | - | 303 | ||||
AK008681.1 | - | 2 | 269 | |||
AK031631.1 | - | 2 | 250 | |||
AK053644.1 | - | 2 | 312 | |||
AK089027.1 | - | 2 | 303 | |||
AK147060.1 | - | 2 | 312 | |||
AK149996.1 | - | 2 | 309 | |||
BC016432.1 | - | 2 | 254 | 4217820 | ||
BC019174.1 | - | 2 | 305 | 5007578 | ||
BC024871.1 | - | 2 | 252 | 3154594 | ||
BC058191.1 | - | 2 | 282 | 5340253 |