Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm21992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290127.1 | NP_001277056.1 | 1 | 374 | ||
NM_001290128.1 | NP_001277057.1 | 1 | 374 | |||
GenBank mRNA | AK044883.1 | - | 354 | |||
AK087488.1 | - | 1 | 344 | |||
EU287940.1 | - | 1 | 248 | |||
EU287941.1 | - | 1 | 248 |
Gene Symbol: Rbm14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019869.3 | NP_063922.2 | 1 | 374 | ||
GenBank mRNA | AK004928.1 | - | 1 | 344 | ||
AK050945.1 | - | 1 | 376 | |||
AK088201.1 | - | 1 | 340 | |||
AK153230.1 | - | 1 | 349 | |||
AK167503.1 | - | 1 | 353 | |||
BB854526.1 | - | 1 | 375 | |||
BC010294.1 | - | 1 | 325 | 3153900 |