Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027204.2 | NP_081480.2 | 5 | 826 | ||
GenBank mRNA | AK002757.1 | - | 5 | 836 | ||
AK005328.1 | - | 5 | 857 | |||
AK028168.1 | - | 5 | 839 | |||
AK134816.1 | - | 5 | 859 | |||
AK166969.1 | - | 5 | 855 | |||
AK168337.1 | - | 5 | 829 | |||
AK205740.1 | - | 1 | 157 | |||
AK212489.1 | - | 1 | 157 | |||
AK219774.1 | - | 1 | 157 | |||
BC005712.1 | - | 5 | 826 | 3582724 | ||
BC019565.1 | - | 5 | 824 | 4219731 |