Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026490.2 | NP_080766.1 | 2 | 294 | ||
GenBank mRNA | AK021180.1 | - | 2 | 288 | ||
AK030793.1 | - | 2 | 282 | |||
AK050637.1 | - | 2 | 293 | |||
AK141554.1 | - | 2 | 304 | |||
BC020315.2 | - | 2 | 270 | |||
BC043921.1 | - | 2 | 275 | 5361515 | ||
BC061064.1 | - | 2 | 278 | 6535226 |