Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019637.3 | NP_062611.2 | 3, 4 | 435 | ||
GenBank mRNA | AK002822.1 | - | 377 | |||
AK030122.1 | - | 3, 4 | 435 | |||
AK046970.1 | - | 3, 4 | 412 | |||
AK154136.1 | - | 3, 4 | 530 | |||
AK166043.1 | - | 3, 4 | 418 | |||
BC038608.1 | - | 3, 4 | 385 | 5012303 | ||
BC085089.1 | - | 3, 4 | 382 | 6492329 | ||
U34973.1 | - | 3, 4 | 403 |