Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019637.3 | NP_062611.2 | 5 | 550 | ||
GenBank mRNA | AK002822.1 | - | 492 | |||
AK030122.1 | - | 5 | 550 | |||
AK042741.1 | - | 6 | 605 | |||
AK046970.1 | - | 5 | 527 | |||
AK154136.1 | - | 5 | 645 | |||
AK166043.1 | - | 5 | 533 | |||
BC038608.1 | - | 5 | 500 | 5012303 | ||
BC085089.1 | - | 5 | 497 | 6492329 | ||
BC096674.1 | - | 4 | 460 | 5144003 | ||
U34973.1 | - | 5 | 518 |