Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177965.1 | NP_001171436.1 | 6 | 531 | ||
NM_019870.3 | NP_063923.1 | 6 | 531 | |||
GenBank mRNA | AJ627039.1 | - | 5 | 350 | ||
AK003806.1 | - | 6 | 468 | |||
AK009697.1 | - | 6 | 442 | |||
AK014469.1 | - | 6 | 460 | |||
AK078700.1 | - | 5 | 701 | |||
AY183136.1 | - | 6 | 500 | |||
AY183137.1 | - | 6 | 500 | |||
BC027219.1 | - | 6 | 432 | 3673739 |
Gene Symbol: Arhgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129060.1 | - | 418 |