Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019871.2 | NP_063924.2 | 2 | 627 | ||
GenBank mRNA | AF016712.1 | - | 2 | 478 | ||
AK077181.1 | - | 2 | 753 | |||
BC137893.1 | - | 2 | 507 | 8860912 | ||
BC137894.1 | - | 2 | 507 | 8860913 |