Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301643.1 | NP_001288572.1 | 10 | 829 | ||
NM_001301644.1 | NP_001288573.1 | 10 | 798 | |||
NM_001301666.1 | NP_001288595.1 | 9 | 745 | |||
NM_019640.5 | NP_062614.1 | 10 | 829 | |||
GenBank mRNA | AK028235.1 | - | 10 | 764 | ||
BC034676.1 | - | 10 | 719 | 4481685 | ||
BQ178694.1 | - | 9 | 634 | |||
U46934.1 | - | 10 | 731 |