Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320198.1 | NP_001307127.1 | 11 | 1886 | ||
XM_005268866.4 | XP_005268923.1 | 11 | 6122 | |||
XM_017019296.1 | XP_016874785.1 | 10 | 1971 | |||
GenBank mRNA | AK292237.1 | - | 11 | 1857 | ||
AK297794.1 | - | 10 | 1811 | |||
BG216771.1 | - | 2 | 434 | |||
X99142.1 | - | 9 | 1667 |