Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019805.4 | NP_062779.3 | 5 | 750 | ||
GenBank mRNA | AF076607.2 | - | 5 | 714 | ||
AK030775.1 | - | 5 | 715 | |||
AK049968.1 | - | 5 | 689 | |||
AK137286.1 | - | 5 | 714 | |||
AK150764.1 | - | 5 | 714 | |||
AK150891.1 | - | 5 | 714 | |||
AK153114.1 | - | 5 | 714 | |||
AK159523.1 | - | 5 | 682 | |||
AK164202.1 | - | 5 | 719 | |||
AK188802.1 | - | 2 | 217 | |||
BC006635.1 | - | 5 | 703 | 3482261 |