Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KTN1 SIRNA Assays ›
Gene Symbol:
KTN1
Gene Name:
kinectin 1
Gene Aliases:
CG1, KNT, MU-RMS-40.19
Chromosome Location:
Chr.14: 55580207 - 55684584 on Build GRCh38
UniGene ID:
Hs.509414
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8034
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KTN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079521.1
NP_001072989.1
37
3677
NM_001079522.1
NP_001072990.1
37
3732
NM_001271014.1
NP_001257943.1
38
3801
NM_004986.3
NP_004977.2
36
3590
NR_073128.1
37
3732
NR_073129.1
38
3801
XM_006720138.2
XP_006720201.1
38
3792
XM_006720139.2
XP_006720202.1
37
3640
XM_006720140.3
XP_006720203.1
36
3553
XM_006720141.2
XP_006720204.1
35
3463
XM_011536751.2
XP_011535053.1
37
3898
XM_011536752.1
XP_011535054.1
36
3571
XM_011536753.1
XP_011535055.1
36
3571
XM_017021278.1
XP_016876767.1
37
9490
XM_017021279.1
XP_016876768.1
35
3484
XM_017021280.1
XP_016876769.1
36
3636
XM_017021281.1
XP_016876770.1
36
9458
XM_017021282.1
XP_016876771.1
37
3708
XM_017021283.1
XP_016876772.1
37
3642
XM_017021284.1
XP_016876773.1
35
3484
XM_017021285.1
XP_016876774.1
36
3616
XM_017021286.1
XP_016876775.1
34
3394
GenBank mRNA
AK055870.1
-
3
1514
AK125499.1
-
5
1021
AK300703.1
-
6
748
AK302797.1
-
38
3755
AK308272.1
-
19
1410
AY264265.1
-
38
3686
AY536375.1
-
27
1890
BC112337.1
-
37
3615
40068009
BC117132.1
-
36
3549
40125683
BC143720.1
-
37
3618
9052234
BX537639.1
-
1
531
D13629.1
-
36
3495
L25616.1
-
35
3452
Z22551.1
-
37
3565
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
cell-cell adhesion
protein binding
kinesin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline