Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019874.3 | NP_063927.1 | 2 | 745 | ||
GenBank mRNA | AF088983.1 | - | 3 | 792 | ||
AF321322.1 | - | 3 | 785 | |||
AK158535.1 | - | 3 | 837 | |||
AK158972.1 | - | 3 | 1045 | |||
BC027782.1 | - | 2 | 3488 | |||
BC048902.1 | - | 3 | 1030 | 6396037 | ||
BC057087.1 | - | 2 | 745 | 5695243 |