Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019835.2 | NP_062809.2 | 4 | 552 | ||
GenBank mRNA | AB004786.1 | - | 4 | 553 | ||
AF142673.1 | - | 4 | 406 | |||
AK151469.1 | - | 1 | 22 | |||
AK152652.1 | - | 1 | 22 | |||
BE981379.1 | - | 4 | 422 | |||
CJ134591.1 | - | 4 | 553 |