Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047159.2 | NP_001040624.1 | 8 | 1123 | ||
NM_019671.3 | NP_062645.2 | 10 | 1244 | |||
GenBank mRNA | AF094520.1 | - | 10 | 1140 | ||
AJ010045.1 | - | 8 | 1086 | |||
AK082128.1 | - | 8 | 1120 | |||
AK088632.1 | - | 8 | 1087 | |||
AK139921.1 | - | 8 | 1087 | |||
BC004699.1 | - | 8 | 1097 | 3499258 | ||
CA980605.1 | - | 4 | 426 | |||
CN697204.1 | - | 2 | 243 |