Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019766.4 | NP_062740.1 | 6 | 713 | ||
GenBank mRNA | AB024935.1 | - | 6 | 521 | ||
AF153479.1 | - | 5 | 410 | |||
AK008805.1 | - | 6 | 744 | |||
AK017742.1 | - | 6 | 531 | |||
AK075932.1 | - | 6 | 578 | |||
AK075987.1 | - | 6 | 624 | |||
AK077538.1 | - | 6 | 714 | |||
AK167443.1 | - | 6 | 689 | |||
AK168073.1 | - | 6 | 624 | |||
AK168210.1 | - | 6 | 624 | |||
AK168721.1 | - | 6 | 563 | |||
AY281130.1 | - | 6 | 740 | |||
BC003708.1 | - | 6 | 606 | 3489418 | ||
BC085264.1 | - | 6 | 610 | 6392001 |