Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019766.4 | NP_062740.1 | 4 | 519 | ||
GenBank mRNA | AB024935.1 | - | 4 | 327 | ||
AF153479.1 | - | 3 | 216 | |||
AK008805.1 | - | 4 | 550 | |||
AK010636.1 | - | 4 | 521 | |||
AK017742.1 | - | 4 | 337 | |||
AK075932.1 | - | 4 | 384 | |||
AK075987.1 | - | 4 | 430 | |||
AK077538.1 | - | 4 | 520 | |||
AK167443.1 | - | 4 | 495 | |||
AK168073.1 | - | 4 | 430 | |||
AK168210.1 | - | 4 | 430 | |||
AK168721.1 | - | 4 | 369 | |||
AY281130.1 | - | 4 | 546 | |||
BC003708.1 | - | 4 | 412 | 3489418 | ||
BC085264.1 | - | 4 | 416 | 6392001 |