Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019877.2 | NP_063930.1 | 3 | 230 | ||
GenBank mRNA | AB040137.1 | - | 3 | 233 | ||
AB047850.1 | - | 3 | 243 | |||
AF237687.1 | - | 3 | 233 | |||
AK003622.1 | - | 3 | 153 | |||
AK187566.1 | - | 2 | 112 | |||
BC025122.1 | - | 3 | 219 | 4187541 |