Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019701.2 | NP_062675.2 | 7 | 640 | ||
GenBank mRNA | AF124847.1 | - | 7 | 633 | ||
AK146876.1 | - | 6 | 703 | |||
BC019983.1 | - | 7 | 666 | 4188062 |