Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039137.3 | NP_001034226.1 | 3 | 249 | ||
NM_001285992.1 | NP_001272921.1 | 3 | 351 | |||
NM_019708.4 | NP_062682.1 | 3 | 354 | |||
GenBank mRNA | AF115778.1 | - | 3 | 172 | ||
AK002521.1 | - | 3 | 203 | |||
AK003095.1 | - | 3 | 197 | |||
AK076085.1 | - | 3 | 147 | |||
AK088272.1 | - | 3 | 187 | |||
AK167488.1 | - | 3 | 197 | |||
AK207171.1 | - | 1 | 30 | |||
BC017629.1 | - | 3 | 173 | |||
BC053746.1 | - | 2 | 3080 | |||
BC085265.1 | - | 3 | 161 | 6391098 | ||
BC092071.1 | - | 3 | 157 | 4505111 | ||
BC108407.1 | - | 3 | 171 | |||
BC147069.1 | - | 3 | 242 | 9056075 | ||
BY251609.1 | - | 3 | 355 | |||
CX241959.1 | - | 3 | 340 |