Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029565.3 | NP_083841.4 | 8 | 947 | ||
GenBank mRNA | AK003252.1 | - | 8 | 861 | ||
AK050162.1 | - | 8 | 865 | |||
AK145644.1 | - | 8 | 868 | |||
AK159761.1 | - | 8 | 875 | |||
AK166772.1 | - | 8 | 862 | |||
AK167058.1 | - | 8 | 861 | |||
AK177231.1 | - | 4 | 280 | |||
AX775905.1 | - | 8 | 882 | |||
BC002164.1 | - | 2 | 126 | |||
BC014732.1 | - | 8 | 851 | 4235248 | ||
BC018379.1 | - | 8 | 869 | 4242583 | ||
BC045145.1 | - | 8 | 885 | 6475115 | ||
BC058273.1 | - | 8 | 862 | 6494379 |