Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029565.3 | NP_083841.4 | 2 | 363 | ||
GenBank mRNA | AF116911.1 | - | 2 | 314 | ||
AK003252.1 | - | 2 | 277 | |||
AK050162.1 | - | 2 | 281 | |||
AK145644.1 | - | 2 | 284 | |||
AK159761.1 | - | 2 | 291 | |||
AK166772.1 | - | 2 | 278 | |||
AK167058.1 | - | 2 | 277 | |||
AK204157.1 | - | 2 | 96 | |||
AX775905.1 | - | 2 | 298 | |||
BB860994.1 | - | 2 | 363 | |||
BC014732.1 | - | 2 | 267 | 4235248 | ||
BC018379.1 | - | 2 | 285 | 4242583 | ||
BC045145.1 | - | 2 | 301 | 6475115 | ||
BC058273.1 | - | 2 | 278 | 6494379 |