Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285793.1 | NP_001272722.1 | 3, 4 | 815 | ||
NM_019804.4 | NP_062778.2 | 3, 4 | 748 | |||
GenBank mRNA | AF142672.1 | - | 3, 4 | 725 | ||
AF158746.1 | - | 3, 4 | 727 | |||
AK018658.1 | - | 3, 4 | 712 | |||
AK041692.1 | - | 3, 4 | 740 | |||
AK045602.1 | - | 3, 4 | 720 | |||
AK050005.1 | - | 3, 4 | 744 | |||
AK052013.1 | - | 3, 4 | 742 | |||
AK053603.1 | - | 3, 4 | 809 | |||
AK085368.1 | - | 3, 4 | 734 | |||
AK136637.1 | - | 3, 4 | 660 | |||
AK188220.1 | - | 1, 2 | 127 | |||
BC013492.1 | - | 3, 4 | 711 | 4218187 | ||
BC031115.1 | - | 3, 4 | 800 | 5026168 |