Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285793.1 | NP_001272722.1 | 3 | 596 | ||
NM_019804.4 | NP_062778.2 | 3 | 529 | |||
GenBank mRNA | AF142672.1 | - | 3 | 506 | ||
AF158746.1 | - | 3 | 508 | |||
AK018658.1 | - | 3 | 493 | |||
AK041692.1 | - | 3 | 521 | |||
AK045602.1 | - | 3 | 501 | |||
AK050005.1 | - | 3 | 525 | |||
AK052013.1 | - | 3 | 523 | |||
AK053603.1 | - | 3 | 590 | |||
AK085368.1 | - | 3 | 515 | |||
AK136637.1 | - | 3 | 441 | |||
BC013492.1 | - | 3 | 492 | 4218187 | ||
BC031115.1 | - | 3 | 581 | 5026168 |