Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190852.1 | NP_001177781.1 | 6 | 622 | ||
NM_001190853.1 | NP_001177782.1 | 8 | 580 | |||
NM_001190854.1 | NP_001177783.1 | 6 | 622 | |||
NM_001190855.1 | NP_001177784.1 | 6 | 622 | |||
NM_001190856.1 | NP_001177785.1 | 6 | 622 | |||
NM_001190857.1 | NP_001177786.1 | 6 | 529 | |||
NM_019808.3 | NP_062782.2 | 6 | 949 | |||
NM_019809.3 | NP_062783.2 | 6 | 949 | |||
NM_022554.3 | NP_072048.2 | 7 | 544 | |||
GenBank mRNA | AA061806.1 | - | 6 | 266 | ||
AA512794.1 | - | 6 | 289 | |||
AB016586.1 | - | 6 | 871 | |||
AB016587.1 | - | 7 | 556 | |||
AB016588.1 | - | 6 | 871 | |||
AI391027.1 | - | 2 | 415 | |||
AK003194.1 | - | 8 | 523 | |||
AK009464.1 | - | 8 | 492 | |||
AK089779.1 | - | 6 | 953 | |||
AK147999.1 | - | 6 | 889 | |||
BC037476.1 | - | 6 | 913 | 4457868 | ||
CB597177.1 | - | 8 | 586 | |||
DQ177283.1 | - | 5 | 796 | |||
HM369794.1 | - | 5 | 469 | |||
HM369795.1 | - | 5 | 469 | |||
HM369796.1 | - | 5 | 469 | |||
HM369797.1 | - | 5 | 469 | |||
HM369798.1 | - | 6 | 487 |