Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019689.2 | NP_062663.1 | 5 | 969 | ||
GenBank mRNA | AF116847.1 | - | 5 | 969 | ||
AK084952.1 | - | 3 | 281 | |||
AK139455.1 | - | 7 | 1151 | |||
AK148073.1 | - | 6 | 1066 | |||
BC145330.1 | - | 5 | 1033 | |||
BC150755.1 | - | 5 | 963 | 9087666 |