Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019773.2 | NP_062747.1 | 3 | 829 | ||
GenBank mRNA | AB027290.1 | - | 3 | 658 | ||
AB232602.1 | - | 1 | 443 | |||
AK010710.1 | - | 3 | 608 | |||
AK017301.1 | - | 3 | 623 | |||
AK032133.1 | - | 3 | 594 | |||
BC008160.1 | - | 3 | 628 | 3964134 |