Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019694.1 | NP_062668.1 | 6 | 1075 | ||
GenBank mRNA | AB041582.1 | - | 6 | 1088 | ||
AF061026.1 | - | 6 | 1075 | |||
AK133849.1 | - | 6 | 1094 | |||
AK206007.1 | - | 1 | 68 | |||
AK208931.1 | - | 1 | 51 | |||
AK213040.1 | - | 1 | 47 | |||
BC023862.1 | - | 6 | 1072 | |||
BC046326.1 | - | 6 | 1082 | |||
BC055865.1 | - | 6 | 1093 | |||
BC061115.1 | - | 6 | 1088 | 6533034 | ||
BC061690.1 | - | 6 | 1075 |