Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019694.1 | NP_062668.1 | 13 | 2199 | ||
GenBank mRNA | AB041582.1 | - | 13 | 2212 | ||
AF061026.1 | - | 13 | 2199 | |||
AK133849.1 | - | 13 | 2219 | |||
AK161638.1 | - | 6 | 2958 | |||
AK193054.1 | - | 2 | 231 | |||
AK199956.1 | - | 2 | 221 | |||
AK206662.1 | - | 2 | 239 | |||
AK207004.1 | - | 3 | 326 | |||
AK210262.1 | - | 2 | 254 | |||
AK210772.1 | - | 1 | 73 | |||
AK215386.1 | - | 2 | 280 | |||
AK217209.1 | - | 2 | 251 | |||
BC023862.1 | - | 13 | 2196 | |||
BC046326.1 | - | 13 | 2206 | |||
BC055865.1 | - | 13 | 2217 | |||
BC061115.1 | - | 13 | 2212 | 6533034 | ||
BC061690.1 | - | 13 | 2198 |