Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019737.2 | NP_062711.1 | 6 | 1651 | ||
GenBank mRNA | AF097158.1 | - | 6 | 1649 | ||
AK137385.1 | - | 6 | 1631 | |||
AK138247.1 | - | 6 | 1549 | |||
AK138610.1 | - | 6 | 1633 | |||
AK144997.1 | - | 6 | 1525 | |||
AK163956.1 | - | 6 | 1590 | |||
AK164262.1 | - | 6 | 1590 | |||
AK164724.1 | - | 6 | 1590 | |||
BC011149.1 | - | 6 | 874 | 4037384 |