Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019792.2 | NP_062766.2 | 2 | 228 | ||
GenBank mRNA | AF204959.1 | - | 2 | 227 | ||
AK149541.1 | - | 2 | 230 | |||
BC028855.1 | - | 2 | 219 | 5124151 | ||
BY099368.1 | - | 2 | 230 | |||
Y11995.1 | - | 2 | 230 |