Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168505.1 | NP_001161977.1 | 6, 7 | 1617 | ||
NM_019658.6 | NP_062632.2 | 6, 7 | 1658 | |||
GenBank mRNA | AF068921.1 | - | 5, 6 | 1293 | ||
AK146447.1 | - | 6, 7 | 1655 | |||
BC013722.1 | - | 2, 3 | 193 | |||
BC049775.1 | - | 5, 6 | 1523 | 6515905 | ||
BC083060.1 | - | 6, 7 | 1617 | 5356514 |