Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019704.2 | NP_062678.1 | 1 | 965 | ||
GenBank mRNA | AF134238.1 | - | 1 | 965 | ||
AK043834.1 | - | 1 | 893 | |||
AK154202.1 | - | 1 | 890 | |||
AK170756.1 | - | 1 | 824 | |||
AK170824.1 | - | 1 | 853 | |||
BC019473.1 | - | 1 | 821 | 4161444 |