Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morf4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168225.1 | NP_001161697.1 | 4 | 853 | ||
NM_001168226.1 | NP_001161698.1 | 4 | 786 | |||
NM_001168227.1 | NP_001161699.1 | 4 | 738 | |||
NM_001168228.1 | NP_001161700.1 | 4 | 737 | |||
NM_001168229.1 | NP_001161701.1 | 3 | 709 | |||
NM_001168230.1 | NP_001161702.1 | 3 | 647 | |||
NM_019768.4 | NP_062742.4 | 4 | 860 | |||
GenBank mRNA | AB025049.1 | - | 2 | 548 | ||
AK075946.1 | - | 4 | 713 | |||
AK088849.1 | - | 1 | 567 | |||
AK089963.1 | - | 4 | 714 | |||
AK129038.1 | - | 4 | 694 | |||
AK131944.1 | - | 4 | 713 | |||
AK145686.1 | - | 4 | 713 | |||
AK150401.1 | - | 4 | 713 | |||
AK150670.1 | - | 4 | 713 | |||
AK151570.1 | - | 4 | 713 | |||
AK153146.1 | - | 4 | 709 | |||
AK159145.1 | - | 4 | 762 | |||
AK159179.1 | - | 4 | 713 | |||
AK160012.1 | - | 4 | 713 | |||
AK161610.1 | - | 4 | 739 | |||
AK164433.1 | - | 4 | 713 | |||
AK165985.1 | - | 4 | 713 | |||
AK166637.1 | - | 4 | 739 | |||
AK178597.1 | - | 1 | 236 | |||
AK199067.1 | - | 1 | 236 | |||
BC075653.1 | - | 4 | 860 | 30539330 | ||
BC088731.1 | - | 1 | 494 | 5340043 |